Laemmli SDS sample buffer, reducing (6X)
Laemmli SDS Sample Buffer, reducing, 6X, 50 mL
Anbieter: Thermo Fisher Scientific
Laemmli SDS sample buffer, reducing (6X)
Katalognr. J61337.AD
Anbieter: Thermo Fisher Scientific
CAS Nummer:
Direkt zu
Product Details & Documents
Physical Form: Liquid
Appearance: Blue
Applications: For protein sample preparation to be used in the Laemmli SDS-PAGE system
Documents
Technische Daten
Technische Daten
- Pk:50 mL
- Description:Laemmli SDS Sample Buffer, reducing, 6X
- Cat. No.:J61337.AD
- Physical form:Liquid
- Appearance:Blue



