Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Phosphate, 0,2 M buffer solution, pH 4,4

Phosphate buffer soln, 0,2 M, 4,4, 500 mL
Catalog # J60467.AP
Phosphate, 0,2 M buffer solution, pH 4,4
Catalog # J60467.AP
CAS Number:  
Phosphate, 0,2 M buffer solution, pH 4,4
Phosphate, 0,2 M buffer solution, pH 4,4

Phosphate buffer soln, 0,2 M, 4,4, 500 mL

CAS Number:  
Price
123.00 SFr.
Log in for availability
1 * 500 mL
Product Details & Documents

CAS No.: 7558-80-7
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
MDL No.: MFCD00003527

Specifications

Specifications

  • Concentration:
    0,2 M
  • Pk:
    500 mL
  • Description:
    Phosphate buffer soln
  • pH value:
    4,4
  • Cat. No.:
    J60467.AP
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00003527
  • CAS No.:
    7558-80-7

Specifications