Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Stripping buffer-2 (4X)

Stripping buffer-2 (4X)

Stripping buffer, 4X, 250 mL
Catalog # J62023.AK
Stripping buffer-2 (4X)
Catalog # J62023.AK
CAS Number:  
Stripping buffer-2 (4X)
Stripping buffer-2 (4X)

Stripping buffer, 4X, 250 mL

CAS Number:  
Price
SFr. 81.00
Log in for availability
1 * 250 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Stripping buffer, 4X
  • Cat. No.:
    J62023.AK
  • Physical form:
    Liquid

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.