Order Entry
Switzerland
Orders LinkContactUsLinkComponent
GTE buffer

GTE buffer

250 mL
Catalog # J62597.AK
GTE buffer
Catalog # J62597.AK
CAS Number:  
GTE buffer
GTE buffer

250 mL

CAS Number:  
Price
106.00 SFr.
Log in for availability
1 * 250 mL
Product Details & Documents

Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Applications: For miniprep and DNA purification

Specifications

Specifications

  • Pk:
    250 mL
  • Cat. No.:
    J62597.AK
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.