Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Nitrocellulose stripping buffer (10X)

Nitrocellulose stripping buffer (10X)

Nitrocellulose Stripping Buffer, 10X, 250 mL
Catalog # J62541.AK
Nitrocellulose stripping buffer (10X)
Catalog # J62541.AK
CAS Number:  
Nitrocellulose stripping buffer (10X)
Nitrocellulose stripping buffer (10X)

Nitrocellulose Stripping Buffer, 10X, 250 mL

CAS Number:  
Price
SFr. 114.00
Log in for availability
1 * 250 mL
Product Details & Documents

Physical Form: Liquid

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Nitrocellulose Stripping Buffer, 10X
  • Cat. No.:
    J62541.AK
  • Physical form:
    Liquid

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.