Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Thioflavin T ≥75%, tech.

Thioflavin T ≥75%, tech.

25 g
Catalog # J61043.14
Thioflavin T ≥75%, tech.
Catalog # J61043.14
CAS Number:  2390-54-7
Thioflavin T ≥75%, tech.
Thioflavin T ≥75%, tech.

25 g

CAS Number:  2390-54-7
Price
SFr. 137.00
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 75% CAS No.: 2390-54-7 Molecular Formula: C17H19ClN2S Formula Weight: 318.86 Physical Form: Powder Appearance: Yellow Beilstein Reference: 3922452 MDL No.: MFCD00011944

Applications: A dye used to visualize beta-amyloid plaques, like those associated with Alzheimer's Disease

Synonyms:  CI 49005, Thioflavine T, Basic Yellow 1, 2-(4-(Dimethylamino)phenyl)-3,6-dimethylbenzo[d]thiazol-3-ium chloride, Setoflavine T
  • CAS Number:2390-54-7
  • Formula:C₁₇H₁₉ClN₂S
  • MDL Number:MFCD00011944
  • Molecular Weight:318.86 g/mol
  • Storage Temperature:Ambient
  • EINECS:219-228-9
Specifications

Specifications

  • Pk:
    25 g

Specifications