Thioflavin T ≥75%, tech.
5 g
Supplier: Thermo Fisher Scientific
Thioflavin T ≥75%, tech.
Catalog # J61043.06
Supplier: Thermo Fisher Scientific
CAS Number: 2390-54-7
Jump to:
Product Details & Documents
Purity: 75% CAS No.: 2390-54-7 Molecular Formula: C17H19ClN2S Formula Weight: 318.86 Physical Form: Powder Appearance: Yellow Beilstein Reference: 3922452 MDL No.: MFCD00011944
Applications: A dye used to visualize beta-amyloid plaques, like those associated with Alzheimer's Disease
Synonyms: CI 49005, Thioflavine T, Basic Yellow 1, 2-(4-(Dimethylamino)phenyl)-3,6-dimethylbenzo[d]thiazol-3-ium chloride, Setoflavine T
- CAS Number:2390-54-7
- Formula:C₁₇H₁₉ClN₂S
- MDL Number:MFCD00011944
- Molecular Weight:318.86 g/mol
- Storage Temperature:Ambient
- EINECS:219-228-9
Documents
Specifications
Specifications
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



