Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), Powder packages, 4 L
Supplier: Thermo Fisher Scientific
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Catalog # J62449.K7
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00236358
Documents
Specifications
Specifications
- Pack type:Powder packages
- Pk:4 L
- Description:TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)
- Cat. No.:J62449.K7
- Physical form:Liquid
- MDL No.:MFCD00236358
- Appearance:Colourless
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



