Phosphate, 0,5M buffer solution, pH 8,0
Phosphate buffer soln, 0,5 M, 8,0, 500 mL
Supplier: Thermo Fisher Scientific
Phosphate, 0,5M buffer solution, pH 8,0
Catalog # J61722.AP
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527
Documents
Specifications
Specifications
- Concentration:0,5 M
- Pk:500 mL
- Description:Phosphate buffer soln
- pH value:8,0
- Cat. No.:J61722.AP
- Physical form:Liquid
- MDL No.:MFCD00003527
- CAS No.:7558-80-7
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



