AMPD, 0.2 M Buffer Solutions
AMPD buffer solution, 0,2 M, 9,5, 250 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
2-Amino-2-methyl-1,3-propanediol buffer, colorless and odorless.
CAS No.: 115-69-5
MDL No.: MFCD00004678
Caution: Storage Temperature: 2 to 8 °C
Documents
Specifications
Specifications
- Concentration:0,2 M
- Pk:250 mL
- Description:AMPD buffer solution
- pH value:9,5
- Cat. No.:J60916.AK
- Odour/scent:Odourless
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- MDL No.:MFCD00004678
- Appearance:Colourless
- CAS No.:115-69-5
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



