Order Entry
Switzerland
Orders LinkContactUsLinkComponent
MOPS-SDS running buffer (20X)

MOPS-SDS running buffer (20X)

MOPS-SDS running buffer (20X), 500 mL
Catalog # J62847.AP
MOPS-SDS running buffer (20X)
Catalog # J62847.AP
CAS Number:  
MOPS-SDS running buffer (20X)
MOPS-SDS running buffer (20X)

MOPS-SDS running buffer (20X), 500 mL

CAS Number:  
Price
SFr. 225.00
Log in for availability
1 * 500 mL
Product Details & Documents

Physical Form: Liquid

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    MOPS-SDS running buffer (20X)
  • Cat. No.:
    J62847.AP
  • Physical form:
    Liquid

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.