Amphotericin B
100 mg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Amphotericin B, antifungal activity has been used against leishmaniasis caused by protozoan parasites of the Leishmania genus.
- Moisture and light sensitive
Amphotericin B trihydrate was observed to cause suppression of bone marrow progenitor cells and to induce Actinomycin D(CR001) sensitivity in drug resistant HELA cells in vitro.
Yellow powder
Soluble in aqueous buffers at pH 2.0 or 11.0. Insoluble at pH 6.0-7.0. Also soluble in DMSO or DMF
Synonyms: (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid
- CAS Number:1397-89-3
- Formula:C₄₇H₇₃NO₁₇
- MDL Number:MFCD00877763
- Molecular Weight:924.08 g/mol
- Merck Index:14,585
- Storage Temperature:Refrigerator
- EINECS:215-742-2
Documents
Specifications
Specifications
- Pk:100 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



