Orders LinkContactUsLinkComponent
Test kits, drop count tests
Test kits, drop count tests
Hypochlorite, model CN-HR, 50000 - 150000 mg/l, 1
Catalog # HACH2687200
Supplier:   Hach
Test kits, drop count tests
Catalog # HACH2687200
Supplier:  Hach
CAS Number:  
Test kits, drop count tests
Test kits, drop count tests
Hypochlorite, model CN-HR, 50000 - 150000 mg/l, 1
CAS Number:  
Price
SFr. 378.00
Log in for availability
1 * 1 ST
Product Details & Documents
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with colour indicator
  • Reagents supplied with the kit
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • Pk:
    1
  • Description:
    Hypochlorite, model CN-HR
  • Range:
    50000 - 150000 mg/l
Specifications