Pipette tips
Eppendorf type, crystal tip, bevelled, 0,5 - 10 µl, -, Transparent, Gilson, Eppendorf, Socorex, Nichiryo, 10 racks, 96 each
Catalog # DELT301-10
Supplier: DELTALAB
Jump to:
Product Details & Documents
Specifications
- Packed:10 racks, 96 each
- Sterile:-
- Volume:0,5 - 10 µl
- Seq:04
- Tip style:Eppendorf type, crystal tip, bevelled
- Tip colour:Transparent
- For use with:Gilson, Eppendorf, Socorex, Nichiryo