Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Bulb pipettes, MBL®, two marks, Class AS

20 ml, 0,030 ± ml
Catalog # BIBBPRA630AS
Supplier:   SciLabware
Bulb pipettes, MBL®, two marks, Class AS
Catalog # BIBBPRA630AS
Supplier:  SciLabware
Bulb pipettes, MBL®, two marks, Class AS
Bulb pipettes, MBL®, two marks, Class AS

20 ml, 0,030 ± ml

Price
SFr. 28.50
Log in for availability
1 * 2 ST
Product Details & Documents

Soda lime glass.

  • Calibrated to deliver (TD, Ex)
  • Permanent amber stain marking
  • With +5 s waiting time
  • Features a safety bulb at the top of the pipette tube
  • Printed with batch code for downloadable batch certificate

 : Comply with ISO 648 and DIN 12690

Specifications
  • Capacity:
    20 ml
  • Seq:
    05
  • Tolerance:
    0,030 ± ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.