Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Bulb pipettes, MBL®, two marks, Class AS

Bulb pipettes, MBL®, two marks, Class AS

1 ml, 0,008 ± ml
Catalog # BIBBPRA620AS
Supplier:   SciLabware
Bulb pipettes, MBL®, two marks, Class AS
Catalog # BIBBPRA620AS
Supplier:  SciLabware
Bulb pipettes, MBL®, two marks, Class AS
Bulb pipettes, MBL®, two marks, Class AS

1 ml, 0,008 ± ml

Price
SFr. 18.90
Log in for availability
1 * 2 ST
Product Details & Documents

Soda lime glass.

  • Calibrated to deliver (TD, Ex)
  • Permanent amber stain marking
  • With +5 s waiting time
  • Features a safety bulb at the top of the pipette tube
  • Printed with batch code for downloadable batch certificate

 : Comply with ISO 648 and DIN 12690

Specifications
  • Capacity:
    1 ml
  • Seq:
    01
  • Tolerance:
    0,008 ± ml