ContactUsLinkComponent
PTFE conforming to FDA and USP Class VI requirements. Alnico V magnet. Dimensions: Ø14mm x 100mm stirrer bar with a Ø15mm x 2mm fixed ring 1 * 1 items
Catalog # COWI7646
Supplier:  COWIE
PTFE conforming to FDA and USP Class VI requirements. Alnico V magnet. Dimensions: Ø14mm x 100mm stirrer bar with a Ø15mm x 2mm fixed ring 1 * 1 items
Catalog # COWI7646
Supplier:  COWIE
Frequently Bought Together