ContactUsLinkComponent
Sucrose Standard 10% - 1.347824 (± 0.00004) manufactured in accordance with ICUMSA guidelines, tested and certified to 2 decimals pl ace. Extended shelf-life of 12 weeks 1 * 15 mL
Catalog # REGEBS10
Supplier:  REAGECON
Sucrose Standard 10% - 1.347824 (± 0.00004) manufactured in accordance with ICUMSA guidelines, tested and certified to 2 decimals pl ace. Extended shelf-life of 12 weeks 1 * 15 mL
Catalog # REGEBS10
Supplier:  REAGECON
Frequently Bought Together