Orders LinkContactUsLinkComponent
Stools
Stool 2, black, made from stamskin top synthetic leather upholstery, 460 - 630 mm, Castors, —
  BIMO9468H-2571-807
 :   bimos
Stools
  BIMO9468H-2571-807
 :  bimos
Stools
Stools
Stool 2, black, made from stamskin top synthetic leather upholstery, 460 - 630 mm, Castors, —
SFr. 202.00
1 * 1 ST
Production stools with Extra large 40 cm Ø seats.
  • Hardwearing stools provide optimum support in industrial and workshop applications
  • Sturdy steel 5 star base
  • Encourage the posture in keeping with the natural S-shape of the spine
  • Seat height adjustment based on pneumatic spring system
ESD standing rest and ESD stools:
Pneumatic spring height adjustment system
With conductive fabric or synthetic leather upholstery
Conductive castors or glides (stools)
Cleanroom stools:
Base frame made from die-cast aluminium, brilliantly polished
Conductive castors/glides for hard floors
Synthetic leather cover with conductive and non-slip properties
Laboratory Stools:
The new stool system from Bimos is a comfortable and flexible solution for short periods of sitting
Its configuration options make the stool system a versatile seating solution for industrial applications, the service sector, healthcare settings, as well as laboratories
Easy ring control
Coloured ring in various colours
Polished aluminium base
Resistant to desinfectants
  • Description:
    Stool 2, black, made from stamskin top synthetic leather upholstery
  • Height:
    460 - 630 mm
  • Seq:
    7
  • Foot style:
    Castors
  • Foot ring:
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..