Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Measuring scoops

Measuring scoops

150 ml, 250 mm, Natural
Catalog # HIRS7780105
Supplier:   HIRSCHMANN
Measuring scoops
Catalog # HIRS7780105
Supplier:  HIRSCHMANN
Measuring scoops

150 ml, 250 mm, Natural

Price
39.00 SFr.
Log in for availability
1 * 12 ST
Product Details & Documents

HDPE, natural colour.

Specifications
  • Capacity:
    150 ml
  • Colour:
    Natural
  • Length:
    250 mm
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.