Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Measuring scoops

Measuring scoops

350 ml, 310 mm, Natural
Catalog # HIRS7780106
Supplier:   HIRSCHMANN
Measuring scoops
Catalog # HIRS7780106
Supplier:  HIRSCHMANN
Measuring scoops

350 ml, 310 mm, Natural

Price
SFr. 36.30
Log in for availability
1 * 6 ST
Product Details & Documents

HDPE, natural colour.

Specifications
  • Capacity:
    350 ml
  • Colour:
    Natural
  • Length:
    310 mm
  • Seq:
    06
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.