ContactUsLinkComponent
Standard rack for upright freezer for (hxd) 3x3 = 9 boxes 130mm high, delivered including standard white cardboard boxes and 9x9 cel l dividers, stainless steel, dimensions (hxdxw) 401x420x140 1 * 1 items
Catalog # TENATE23332B
Supplier:  TENAK
Standard rack for upright freezer for (hxd) 3x3 = 9 boxes 130mm high, delivered including standard white cardboard boxes and 9x9 cel l dividers, stainless steel, dimensions (hxdxw) 401x420x140 1 * 1 items
Catalog # TENATE23332B
Supplier:  TENAK
Frequently Bought Together