ContactUsLinkComponent
Petri plate, round, material PS natural, bottom diameter 88mm, lid diameter 92mm, vents 3, aseptic, height 16.2mm. 1 * 700 items
Catalog # VWRI391-0440_B
Supplier:  VWR Collection
Petri plate, round, material PS natural, bottom diameter 88mm, lid diameter 92mm, vents 3, aseptic, height 16.2mm. 1 * 700 items
Catalog # VWRI391-0440_B
Supplier:  VWR Collection