Orders LinkContactUsLinkComponent
ClipTip™ Racks for Tips
ClipTip™ Racks for Tips
Empty rack, small, colour code pink and purple (for ClipTip 20 and 50)
Catalog # 613-2227
   Low Manufacturing Impact
ClipTip™ Racks for Tips
Catalog # 613-2227
   Low Manufacturing Impact
ClipTip™ Racks for Tips
ClipTip™ Racks for Tips
Empty rack, small, colour code pink and purple (for ClipTip 20 and 50)
Price
22.00 SFr.
Sale
Valid until 30/06/2026
Log in for availability
1 * 10 ST
Product Details & Documents
Utilize Thermo Scientific™ ClipTip™ Pipette tip Racks, available in three sizes.
  • Small rack: For use with ClipTip™ 20 and 50 Pipette tips
  • Medium rack: For use with ClipTip™ 200 and 300 Pipette tips
  • Large rack: For use with ClipTip™ 1000 Pipette tips
Ordering information: Racks are supplied without tips and inserts.
Specifications
  • Description:
    Empty rack, small, colour code pink and purple (for ClipTip 20 and 50)
  • Seq:
    04
  • Cat. No.:
    613-2227
  • Environmental certification:
    ISO 14001 (Certificate Registration No. EMS 623400)
  • Environmentally Preferable:
    Low Manufacturing Impact