Clear inserts, polyester (PET) membrane, Transwell®
24-well insert plates, 0,33 cm², Polyester, 6,5 mm, 8,0 µm
Supplier: Corning
Clear inserts, polyester (PET) membrane, Transwell®
Catalog # 734-2607
Supplier: Corning
Jump to:
Product Details & Documents
Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.
- w Produce a cell culture environment that closely resembles an in vivo state
- Co-culture cells with or without cell-to-cell contact
- Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 µm transparent membrane
- Treated for optimal cell attachment
- Excellent visibility under phase contrast microscopy
- Supplied in multiple well plates
- Sterile
Allow polarised cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally.
Specifications
- Membrane:Polyester
- Pore size:8,0 µm
- Description:24-well insert plates
- Seq:01
- Culture area:0,33 cm²
- Membrane Ø:6,5 mm
- Cat. No.:734-2607
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Environmentally Preferable:Low Manufacturing Impact



