Orders LinkContactUsLinkComponent
VWR®, N-Plan Objectives
VWR®, N-Plan Objectives
IOS N-plan achromatic objective
N° de catalogue 630-1640
Fournisseur:   VWR Collection
VWR®, N-Plan Objectives
N° de catalogue 630-1640
Fournisseur:  VWR Collection
VWR®, N-Plan Objectives
VWR®, N-Plan Objectives
IOS N-plan achromatic objective
Prix
49.30 SFr.
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
These objectives are ideal for use in the education field and for laboratory routine applications.
IOS N-Plan objectives feature an infinity-corrected optical system.
Documents
Spécifications
  • Magnification:
    10×/0,25 mm
  • Description:
    IOS N-plan achromatic objective
  • Seq:
    202