Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

VWR®, N-Plan Objectives

IOS N-plan achromatic objective (oil/water), 0,13 mm
  630-1643
VWR®, N-Plan Objectives
  630-1643
VWR®, N-Plan Objectives
VWR®, N-Plan Objectives

IOS N-plan achromatic objective (oil/water), 0,13 mm

 
SFr. 182.00
   
1 * 1 ST
 

These objectives are ideal for use in the education field and for laboratory routine applications.

IOS N-Plan objectives feature an infinity-corrected optical system.

 
  • Magnification:
    100×/ 1,25 mm (i)
  • Description:
    IOS N-plan achromatic objective (oil/water)
  • Seq:
    206
  • Working distance:
    0,13 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.