Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Copper(I) cyanide

Copper(I) cyanide

2x2,5 kg, 2
Catalog # 12135.F3
Toxic
Environment
Copper(I) cyanide
Catalog # 12135.F3
CAS Number:  544-92-3
Copper(I) cyanide
Copper(I) cyanide

2x2,5 kg, 2

CAS Number:  544-92-3
Price
SFr. 704.00
Log in for availability
1 * 2 ST
Product Details & Documents

CAS No.: 544-92-3 Molecular Formula: CCuN Formula Weight: 89.56 Physical Form: Powder Density: 2.92 Appearance: White to cream Odour: Characteristic Melting Point: 473° Solubility: Practically insoluble in water, alcohol. Soluble in NH4OH Beilstein Reference: 3587244 Merck Reference: 14,2661 UN No.: 1587 MDL No.: MFCD00010975

Electroplating Cu or Fe, polymerization catalyst

Synonyms:  Copper cyanide, Cuprous cyanide
  • CAS Number:544-92-3
  • Formula:CCuN
  • MDL Number:MFCD00010975
  • Molecular Weight:89.56 g/mol
  • Merck Index:14,2661
  • Melting Pt:473 °C
  • Density:2.92
  • Storage Temperature:Ambient
  • EINECS:208-883-6
  • UN:1587
  • ADR:6.1,II
Specifications

Specifications

  • Pk:
    2
  • Pk Info:
    2x2,5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.