ContactUsLinkComponent
Reusable cooling core for use in CoolBox XT and CoolBox 2XT systems freeze in -20 C freezer, use for up to 18 hours of benchtop c ooling 1 * 1 items
Catalog # 216-0953
Supplier:  BIOCISION
undefined
Reusable cooling core for use in CoolBox XT and CoolBox 2XT systems freeze in -20 C freezer, use for up to 18 hours of benchtop c ooling 1 * 1 items
Catalog # 216-0953
Supplier:  BIOCISION
Frequently Bought Together