Cooling elements and frames
Cooling frames, 4 °C, Tempshell, Blue, 2...8 °C
Codice catalogo DLTTA100030DE
Nome del fornitore: DELTA T
Vai a
Product Details & Documents
Molecular medical elements for different temperature handling of samples ranges from −32 °C to +38 °C.
- Made of PE and natural in colour
- Sealing made of laminated aluminium film with a protected cap
- A liquid crystal thermometer in the elements shows its current temperature
Used for transportation and storage of samples like blood products, homograft’s, cell cultures, stem cells, antibodies, proteins and other temperature sensitive medications.
Inside dimension: 210×110×30 mm
Specifiche
- Temperature Range:2...8 °C
- Colour:Blue
- Description:Cooling frames, 4 °C, Tempshell
- Seq:10
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



