Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Heating tapes

Heating tapes

Heating tape, 0,9 m length, 50 W
Catalog # WINKWBS00200900050
Supplier:   WINKLER
Heating tapes
Catalog # WINKWBS00200900050
Supplier:  WINKLER
Heating tapes
Heating tapes

Heating tape, 0,9 m length, 50 W

Price
SFr. 132.00
Log in for availability
Product Details & Documents

These heating tapes contain four heat conductors, insulated silicone, outer braiding with tinned copper wire, factory terminated and vulcanised main connection.

  • Very high and constant heating power per meter
  • High operating temperature
  • Extremely flexible
  • Quick and easy installation
Specifications
  • Description:
    Heating tape, 0,9 m length, 50 W
  • Seq:
    01