Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Heating tapes

Heating tapes

Heating tape, 15,5 m length, 1160 W
Catalog # WINKWBS00215501160
Supplier:   WINKLER
Heating tapes
Catalog # WINKWBS00215501160
Supplier:  WINKLER
Heating tapes
Heating tapes

Heating tape, 15,5 m length, 1160 W

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

These heating tapes contain four heat conductors, insulated silicone, outer braiding with tinned copper wire, factory terminated and vulcanised main connection.

  • Very high and constant heating power per meter
  • High operating temperature
  • Extremely flexible
  • Quick and easy installation
Specifications
  • Description:
    Heating tape, 15,5 m length, 1160 W
  • Seq:
    22
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.