Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Heating tapes

Heating tape, 6,3 m length, 640 W
Catalog # WINKWBS00206300640
Supplier:   WINKLER
Heating tapes
Catalog # WINKWBS00206300640
Supplier:  WINKLER
Heating tapes
Heating tapes

Heating tape, 6,3 m length, 640 W

Price
Inquire For Price
Log in for availability
Product Details & Documents

These heating tapes contain four heat conductors, insulated silicone, outer braiding with tinned copper wire, factory terminated and vulcanised main connection.

  • Very high and constant heating power per meter
  • High operating temperature
  • Extremely flexible
  • Quick and easy installation
Specifications
  • Description:
    Heating tape, 6,3 m length, 640 W
  • Seq:
    15