Heating tapes
Heating tape, 4,0 m length, 410 W
Catalog # WINKWBS00204000410
Supplier: WINKLER
Jump to:
Product Details & Documents
These heating tapes contain four heat conductors, insulated silicone, outer braiding with tinned copper wire, factory terminated and vulcanised main connection.
- Very high and constant heating power per meter
- High operating temperature
- Extremely flexible
- Quick and easy installation
Specifications
- Description:Heating tape, 4,0 m length, 410 W
- Seq:12
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..



