ContactUsLinkComponent
NanoLC-Ultra 2D system. Dual Gradient pump: 1-10 ?l/min for gradient 1, 50-500 nl/min for gradient 2. Max. pressure 10,000 psi. Tem perature controlled column compartment with two 10 port column switching valves. Includes NanoLC start-up kit (p/n 801 1 * 1 items
Catalog # 903-0612
Supplier:  EKSIGENT
NanoLC-Ultra 2D system. Dual Gradient pump: 1-10 ?l/min for gradient 1, 50-500 nl/min for gradient 2. Max. pressure 10,000 psi. Tem perature controlled column compartment with two 10 port column switching valves. Includes NanoLC start-up kit (p/n 801 1 * 1 items
Catalog # 903-0612
Supplier:  EKSIGENT
Frequently Bought Together