ContactUsLinkComponent
NanoLC-Ultra 2D system. Dual Gradient pump: 50-500 nl/min for both gradients. Max. pressure 10,000 psi. Temperature controlled colu mn compartment with one 10 port column switching valve. Includes NanoLC start-up kit (p/n 801-00043) and Eksigent cont 1 * 1 items
Référence 903-0614
Fournisseur:  EKSIGENT
NanoLC-Ultra 2D system. Dual Gradient pump: 50-500 nl/min for both gradients. Max. pressure 10,000 psi. Temperature controlled colu mn compartment with one 10 port column switching valve. Includes NanoLC start-up kit (p/n 801-00043) and Eksigent cont 1 * 1 items
Référence 903-0614
Fournisseur:  EKSIGENT
Frequently Bought Together