ContactUsLinkComponent
Wide mouth bottle with seal cap. Material : high density polyethylene. Color : Neutral – for food. Screw cap with opening-off seal. Recommended for the contents-saves after filling. Suitable the containment of dense fluids, granulates and powders. Em 1 * 58 items
Catalog # LAMA447
Supplier:  LAMAPLAST
Wide mouth bottle with seal cap. Material : high density polyethylene. Color : Neutral – for food. Screw cap with opening-off seal. Recommended for the contents-saves after filling. Suitable the containment of dense fluids, granulates and powders. Em 1 * 58 items
Catalog # LAMA447
Supplier:  LAMAPLAST
Frequently Bought Together