Order Entry
Switzerland
Orders LinkContactUsLinkComponent
(S)-(-)-Nicotine 99+%

(S)-(-)-Nicotine 99+%

Glass bottle, 500 g
Catalog # ACRO181425000
Toxic
Environment
(S)-(-)-Nicotine 99+%
Catalog # ACRO181425000
CAS Number:  54-11-5
(S)-(-)-Nicotine 99+%
(S)-(-)-Nicotine 99+%

Glass bottle, 500 g

CAS Number:  54-11-5
Price
SFr. 764.00
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  (S)-(-)-3-(N-Methyl-2-pyrrolidinyl)pyridine, (S)-(-)-3-(1-Methyl-2-pyrrolidinyl)pyridine, L(-)-Nicotine, L-Nicotine, Nicotine, (-)-Nicotine
  • CAS Number:54-11-5
  • Formula:C₁₀H₁₄N₂
  • MDL Number:MFCD00006369
  • Molecular Weight:162.23 g/mol
  • Merck Index:12,06611
  • Boiling Pt:247 °C (760 mmHg)
  • Melting Pt:–80 °C
  • Density:1.015 g/cm³
  • Flash Pt:101 °C
  • Storage Temperature:Ambient
  • EINECS:200-193-3
  • UN:1654
  • ADR:6.1,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5255 to 1.5285 (20°C, 589 nm)
  • Water:
    ≤0.5 % (K.F.)
  • GC:
    ≥99.0 %
  • Specific optical rotation:
    -140° to -152° (20°C, 589 nm) (c=2,absolute ethanol) (on anhydrous basis)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.