Technical pH buffer solutions, HI 5000 series
8,00, ±0,01, 25 °C, 500 mL
Catalog # HANNHI5008
Supplier: Hanna
Jump to:
Product Details & Documents
- Colour coded labels
- ±0,01 pH
: Technical buffer solutions with certificate.
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:500 mL
- Reference temperature:25 °C
- pH accuracy:±0,01
- pH value:8,00
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



