Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Cryoboxes

-, 15 and 50 ml tubes, White, 145×145×122 mm
  HEATHS2860M
 :   HEATHROW
Cryoboxes
  HEATHS2860M
 :  HEATHROW
Cryoboxes
Cryoboxes

-, 15 and 50 ml tubes, White, 145×145×122 mm

 
SFr. 135.00
   
1 * 10 ST
 

Made of cardboard, white.

  • Write-on surface for descriptions or self numbering systems
  • Dividers are flat packed and comes in two different sizes (to be ordered separately)

For storage of large samples and 15 or 50 ml tubes.

 
  • Compartments:
    -
  • For:
    15 and 50 ml tubes
  • W×D×H ext.:
    145×145×122 mm
  • Colour:
    White
  • Seq:
    01