Cryoboxes
6×6, 15 ml tubes, —, - mm
HEATHS2860T
: HEATHROW
Made of cardboard, white.
- Write-on surface for descriptions or self numbering systems
- Dividers are flat packed and comes in two different sizes (to be ordered separately)
For storage of large samples and 15 or 50 ml tubes.
- Compartments:6×6
- For:15 ml tubes
- W×D×H ext.:- mm
- Colour:—
- Seq:03



