Orders LinkContactUsLinkComponent
Surface profile comparators | Avantor
Surface profile comparators
Surface profile comparator for sand
Katalognr. SIMELD2051
Anbieter:   Industrial Physics
Surface profile comparators
Katalognr. SIMELD2051
Anbieter:  Industrial Physics
Surface profile comparators
Surface profile comparators
Surface profile comparator for sand
Preis
SFr. 358.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Surface comparators for determining roughness by touch and sight.
  • Five different grades of roughness in each comparator
  • Designed to be used with the illuminated magnifier
  • Available in Imperial units only
Section profiles: 0,5 ; 1,0 ; 2,0 ; 3,0 ; 4,0 mils for the three versions.
Zertifizierung: ASTM D4417
Dokumente
Technische Daten
  • Description:
    Surface profile comparator for sand
Accessories
Produktinformationen
  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • SIMELD2054
    Surface profile disc holder and magnifier (5×)
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 151.00