ContactUsLinkComponent
18 COMPOSES A 100 MG/L CHAQUE DANS HNO3 2% Al As B Ba Cd Co Cr Cu Fe Mn Mo Ni Pb Sb Se Ti V Zn 1 * 100 mL
Catalog # 89141.180
18 COMPOSES A 100 MG/L CHAQUE DANS HNO3 2% Al As B Ba Cd Co Cr Cu Fe Mn Mo Ni Pb Sb Se Ti V Zn 1 * 100 mL
Catalog # 89141.180
Frequently Bought Together