Orders LinkContactUsLinkComponent
VWR®, Separating Funnels, Conical
VWR®, Separating Funnels, Conical
100 ml, 19/26 NS
Catalog # 532-0013
Supplier:   VWR Collection
VWR®, Separating Funnels, Conical
Catalog # 532-0013
Supplier:  VWR Collection
VWR®, Separating Funnels, Conical
VWR®, Separating Funnels, Conical
100 ml, 19/26 NS
Price
SFr. 50.70
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate 3.3 glass, with NS-PE stopper.
  • With or without graduations
  • Stopcock with PTFE key
  • Very good chemical and temperature resistance
Specifications
  • Socket size:
    19/26 NS
  • Capacity:
    100 ml
  • Seq:
    08