ekspert microLC 200 system consisting of a binary gradient pumping system configured for 20-200 uL/min flow rate, remotely mounted c olumn oven, electrically-actuated low dispersion 6-port injection valve, and PAL DLW-HTC-xt autosampler with a six-pl 1 * 1 items
Katalognr. 903-0601
Supplier: EKSIGENT
Frequently Bought Together