ContactUsLinkComponent
ekspert microLC 200 systemn consisting of a binary gradient pumping system configured for 20-200 uL/min flow rate, remotely mounted column oven, electrically-actuated low dispersion 6-port injection valve, and PAL DLW-HTC-xt autosampler with a six-pl 1 * 1 items
Catalog # 903-0600
Supplier:  EKSIGENT
undefined
ekspert microLC 200 systemn consisting of a binary gradient pumping system configured for 20-200 uL/min flow rate, remotely mounted column oven, electrically-actuated low dispersion 6-port injection valve, and PAL DLW-HTC-xt autosampler with a six-pl 1 * 1 items
Catalog # 903-0600
Supplier:  EKSIGENT
Frequently Bought Together