Azulene ≥99%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 275-51-4 Molecular Formula: C10H8 Formula Weight: 128.17 Melting Point: 98-101° Boiling Point: 241-243° Merck Reference: 14,926 MDL No.: MFCD00003810
Synonyms: Bicyclo[5.3.0]decapentaene
- CAS Number:275-51-4
- Formula:C₁₀H₈
- MDL Number:MFCD00003810
- Molecular Weight:128.17 g/mol
- Merck Index:14,926
- Boiling Pt:241…243 °C
- Melting Pt:98…101 °C
- Storage Temperature:Ambient
- EINECS:205-993-6
- UN:3077
- ADR:9,III
Documents
Certificates
Search by Lot NumberALFAL08271.03_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



