ContactUsLinkComponent
PRODUCT FOR DNA RELATE APPLICATION, LOAD 6X LOADING DYE ORANGE F, THE 6X LOADING DYE CONTAINS BROMOPHENOL BLUE AND XYLENE CYANOL FF. THE READY-TO-USE SOLUTION CAN BE USED FOR LOADING SAMPLES ON AGAROSE OR POLYACRYLAMIDE GELS. 1 * 6 mL
Catalog # ABNOD0031
Supplier:  Abnova
PRODUCT FOR DNA RELATE APPLICATION, LOAD 6X LOADING DYE ORANGE F, THE 6X LOADING DYE CONTAINS BROMOPHENOL BLUE AND XYLENE CYANOL FF. THE READY-TO-USE SOLUTION CAN BE USED FOR LOADING SAMPLES ON AGAROSE OR POLYACRYLAMIDE GELS. 1 * 6 mL
Catalog # ABNOD0031
Supplier:  Abnova
Frequently Bought Together