ContactUsLinkComponent
FISH PROBES, 9Q SUBTELOMERE(FITC) FISH PROBE, LABELED FISH PROBES FOR IDENTIFICATION OF SUBTELOMERE ABBERRATIONS USING FLUORESCENT I N SITU HYBRIDIZATION TECHNIQUE. 1 * 20 µl
Catalog # ABNOFE0086
Supplier:  Abnova
FISH PROBES, 9Q SUBTELOMERE(FITC) FISH PROBE, LABELED FISH PROBES FOR IDENTIFICATION OF SUBTELOMERE ABBERRATIONS USING FLUORESCENT I N SITU HYBRIDIZATION TECHNIQUE. 1 * 20 µl
Catalog # ABNOFE0086
Supplier:  Abnova
Frequently Bought Together