ContactUsLinkComponent
PRODUCT FOR DNA RELATE APPLICATION, LONGTAQ DNA POLYMERASE, LONGTAQ DNA POLYMERASE CATALYZES 5'-3' SYNTHESIS OF DNA. THE ENZYME HAS BEEN PROVED NOT HAVING THE 3'-5' EXONUCLEASE ACTIVITY. THE ENZYME HAS PROVEN TO HAVE HIGH AMPLIFICATION YIELD, BE STAB 1 * 500 EU
Catalog # ABNOD0013
Supplier:  Abnova
PRODUCT FOR DNA RELATE APPLICATION, LONGTAQ DNA POLYMERASE, LONGTAQ DNA POLYMERASE CATALYZES 5'-3' SYNTHESIS OF DNA. THE ENZYME HAS BEEN PROVED NOT HAVING THE 3'-5' EXONUCLEASE ACTIVITY. THE ENZYME HAS PROVEN TO HAVE HIGH AMPLIFICATION YIELD, BE STAB 1 * 500 EU
Catalog # ABNOD0013
Supplier:  Abnova
Frequently Bought Together